Learn More
R&D Systems™ Recombinant Canine Erythropoietin/EPO Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 3816-CE-010
261.77 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18176870
  • £333.00 / 10µg
Explore more special offers
Description
Description
The Recombinant Canine Erythropoietin Protein is derived from NS0. The Recombinant Canine Erythropoietin Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Erythropoietin/EPO
Mouse myeloma cell line, NS0-derived canine Erythropoietin/EPO protein Ala41-Arg206
ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Recombinant
18.4 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
>90%, by SDS-PAGE under reducing conditions and visualized by silver stain.