missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Syntaxin-6 Protein
A DNA sequence encoding the Gallus gallus (Chicken) Syntaxin-6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6743-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_001006531.1 | |
Syntaxin-6 Protein | |
Research Use Only | |
STX6 | |
Recombinant Protein | |
E. coli | |
EDPFFVVKGE VQKAVNTAQG LFQRWTELLQ DPSIATREEI DWTTNELRNN LRSIEWDLED LDETISIVEA NPRKFNLDAT ELGIRKSFIT STRQVVREMK DQMSNSSMQA LAERKNRQAL LGESSSQSWS SGPDKYSRLD RELQLANSHF IEEQQAQQQL IVEQQDEQLE LVSGSIGVLK NMSQRIGGEL EEQAVMLDDF SHELDSTHSR LDNVMKKLAK VSHMTSDR | |
Greater than 90% as determined by SDS-PAGE. |
424413 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |