missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chikungunya Virus CHIKV E2 Protein

A cDNA sequence encoding the CHIKV E2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11406-100ug

 View more versions of this product

Product Code. 15927168

  • £173.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CHIKV E2 Protein
Research Use Only
Recombinant Protein
E. coli
Sterile Filtered solution containing PBS and 25mM arginine.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Chikungunya Virus
Untagged
Protein is >95% pure as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers