missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chikungunya Virus CHIKV E2 Protein
A cDNA sequence encoding the CHIKV E2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11406-1mg
This item is not returnable.
View return policy
Specifications
CHIKV E2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile Filtered solution containing PBS and 25mM arginine. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chikungunya Virus | |
Untagged | |
Protein is >95% pure as determined by SDS-PAGE. |