missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chlamydia Pneumonia Protein
A cDNA sequence encoding the Chlamydia Pneumonia was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11413-1mg
This item is not returnable.
View return policy
Specifications
Pneumonia Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
PBS buffer and 25mM arginine. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chlamydia | |
His | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |