missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus Lymphotoxin beta R/TNFRSF3 Fc, CF
Carrier-Free Recombinant Protein
Brand: R&D Systems™ 10759-LR-050
311.01 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to inhibit Lymphotoxin alpha 1/beta 2-induced IL-8 secretion in A375 human melanoma cells. Degli-Esposti, M. et al. (1997) J. Immunol. 158:1756. The ED50 for this effect is 30.0-180 ng/mL in the presence of 100 ng/mL Recombinant Human Lymphotoxin alpha 1/beta 2 (Catalog # 8884-LY ).Specifications
XP_005569951.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 57-68 kDa, under reducing conditions, M.W. (Theoretical): 48 kDa | |
Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Lymphotoxin beta R/TNFRSF3 protein Cynomolgus Monkey Lymphotoxin beta R/TNFRSF3 (Ser28-Met227) (N-terminus) Accession # XP_005569951.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
4055 | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
CD18, D12S370, ltbetar, LT-BETA-R, LTBR, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, LymphotoxinbR, TNF RIII, TNF Rrp, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
Reconstitute at 500 μg/mL in PBS. | |
Lymphotoxin beta R/TNFRSF3 |