missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue-2 C-terminal Protein

A cDNA sequence encoding the Dengue-2 C-terminal was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11654-100ug

 View more versions of this product

Product Code. 15957928

  • £173.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Dengue-2 C-terminal Protein
Research Use Only
Recombinant Protein
E. coli
PBS.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
His
Protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers