missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue NS1 ST2 Protein
A cDNA sequence encoding the Dengue NS1 ST2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11645-2ug
This item is not returnable.
View return policy
Specifications
Dengue NS1 ST2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
Dengue NS1 ST2 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, Aprotinin and Pepstatin A. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
His | |
Protein is >95% pure as determined by 12.5% SDS-PAGE. |