missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue Polyvalent Protein

A cDNA sequence encoding the Dengue Polyvalent was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11650-1mg

 View more versions of this product

Product Code. 15947918

  • £1320.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Dengue Polyvalent Protein
Research Use Only
Recombinant Protein
E. coli
Phosphate buffered saline, pH 7.4 and 0.05% NaN3.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers