missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Rapid Test Protein
A cDNA sequence encoding the Dengue Rapid Test was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | Dengue Rapid Test Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Dengue Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927928
|
enQuireBio™
QP11653-100ug |
100 μg |
£173.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15947928
|
enQuireBio™
QP11653-500ug |
500 μg |
£660.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15937928
|
enQuireBio™
QP11653-1mg |
1 mg |
£1320.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Dengue Rapid Test Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
Untagged | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate buffered saline pH 7.4 and 0.02% sodium nitrate. |