missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S10 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8622-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_417780.1 | |
30S ribosomal protein S10 Protein | |
Research Use Only | |
rpsJ | |
Recombinant Protein | |
E. coli | |
MQNQRIRIRL KAFDHRLIDQ ATAEIVETAK RTGAQVRGPI PLPTRKERFT VLISPHVNKD ARDQYEIRTH LRLVDIVEPT EKTVDALMRL DLAAGVDVQI SLG | |
Greater than 90% as determined by SDS-PAGE. |
947816 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |