missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 30S ribosomal protein S10 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8622-ec-100ug

 View more versions of this product

Product Code. 16075621

  • £715.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_417780.1
30S ribosomal protein S10 Protein
Research Use Only
rpsJ
Recombinant Protein
E. coli
MQNQRIRIRL KAFDHRLIDQ ATAEIVETAK RTGAQVRGPI PLPTRKERFT VLISPHVNKD ARDQYEIRTH LRLVDIVEPT EKTVDALMRL DLAAGVDVQI SLG
Greater than 90% as determined by SDS-PAGE.
947816
100 μg
Not determined.
Full Length
E. coli
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers