missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S2 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8615-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_414711.1 | |
30S ribosomal protein S2 Protein | |
Research Use Only | |
rpsB | |
Recombinant Protein | |
E. coli | |
ATVSMRDMLK AGVHFGHQTR YWNPKMKPFI FGARNKVHII NLEKTVPMFN EALAELNKIA SRKGKILFVG TKRAASEAVK DAALSCDQFF VNHRWLGGML TNWKTVRQSI KRLKDLETQS QDGTFDKLTK KEALMRTREL EKLENSLGGI KDMGGLPDAL FVIDADHEHI AIKEANNLGI PVFAIVDTNS DPDGVDFVIP GNDDAIRAVT LYLGAVAATV REGRSQDLAS QAEESFVEAE | |
Greater than 90% as determined by SDS-PAGE. |
947874 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |