missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 4-hydroxy-tetrahydrodipicolinate synthase Protein
A DNA sequence encoding the Escherichia coli (strain K12) 4-hydroxy-tetrahydrodipicolinate synthase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8941-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416973.1 | |
4-hydroxy-tetrahydrodipicolinate synthase Protein | |
Research Use Only | |
dapA | |
Recombinant Protein | |
E. coli | |
MFTGSIVAIV TPMDEKGNVC RASLKKLIDY HVASGTSAIV SVGTTGESAT LNHDEHADVV MMTLDLADGR IPVIAGTGAN ATAEAISLTQ RFNDSGIVGC LTVTPYYNRP SQEGLYQHFK AIAEHTDLPQ ILYNVPSRTG CDLLPETVGR LAKVKNIIGI KEATGNLTRV NQIKELVSDD FVLLSGDDAS ALDFMQLGGH GVISVTANVA ARDMAQMCKL AAEGHFAEAR VINQRLMPLH NKLFVEPNPI PVKWACKELG LVATDTLRLP MTPITDSGRE TVRAALKHAG LL | |
Greater than 90% as determined by SDS-PAGE. |
946952 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |