missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Lipopolysaccharide assembly protein 1
A DNA sequence encoding the Escherichia coli (strain K12) Lipopolysaccharide assembly protein B, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | WP_000891353.1 |
---|---|
Name | Lipopolysaccharide assembly protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | yciM |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16058220
|
enQuireBio™
QP7327-ec-10ug |
10 μg | |||||||
16098220
|
enQuireBio™
QP7327-ec-50ug |
50 μg | |||||||
16048220
|
enQuireBio™
QP7327-ec-100ug |
100 μg |
£715.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16078220
|
enQuireBio™
QP7327-ec-200ug |
200 μg | |||||||
16088220
|
enQuireBio™
QP7327-ec-500ug |
500 μg | |||||||
16068220
|
enQuireBio™
QP7327-ec-1mg |
1 mg |
£2480.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
WP_000891353.1 | |
Research Use Only | |
yciM | |
Recombinant Protein | |
WYMGRRSAQQ NKQDEANRLS RDYVAGVNFL LSNQQDKAVD LFLDMLKEDT GTVEAHLTLG NLFRSRGEVD RAIRIHQTLM ESASLTYEQR LLAIQQLGRD YMAAGLYDRA EDMFNQLTDE TDFRIGALQQ LLQIYQATSE WQKAIDVAER LVKLGKDKQR VEIAHFYCEL ALQHMASDDL DRAMTLLKKG AAADKNSARV SIMMGRVFMA KGEYAKAVES LQRVISQDRE LVSETLEMLQ TCYQQLGKTA EWAEFLQRAV EENTGADAEL MLADIIEARD GSEAAQVYIT RQLQRHPTMR VFHKLMDYHL NEAEEGRAKE SLMVLRDMVG EKVRSKPRYR CQKCGFTAYT LYWHCPSCRA WSTIKPIRGL DGL |
Lipopolysaccharide assembly protein 1 | |
Not determined. | |
Cytoplasmic Domain | |
E. coli | |
Tris-based buffer, 50% glycerol |