missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant E. coli N-Acetyl-D-Glucosamine Kinase/NAGK
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 8020-GK-025
276.45 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to phosphorylate N-acetyl-D-glucosamine. The specific activity is >10,000 pmol/min/μg, as measured under the described conditions.Specifications
P75959 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl, Glycerol, Brij-35 and DTT. | |
M.W. (Observed): 37-38 kDa, reducing conditions; M.W. (theoretical): 34 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
nagK | |
Recombinant | |
N-Acetyl-D-Glucosamine Kinase/NAGK |
Enzyme Activity | |
55577 | |
N-Acetyl-D-Glucosamine Kinase/NAGK | |
E. coli-derived e. coli N-Acetyl-D-Glucosamine Kinase/NAGK protein Met1-Asp303 with C-terminal 6-His tag | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane) |