missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr cytomegalovirus (HCMV) Glycoprotein B / gB Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) cytomegalovirus (HCMV) Glycoprotein B / gB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7275-ec-10ug
This item is not returnable.
View return policy
Specifications
YP_401713.1 | |
cytomegalovirus (HCMV) Glycoprotein B / gB Protein | |
Research Use Only | |
gB | |
Recombinant Protein | |
E. coli | |
QTPEQPAPPA TTVQPTATRQ QTSFPFRVCE LSSHGDLFRF SSDIQCPSFG TRENHTEGLL MVFKDNIIPY SFKVRSYTKI VTNILIYNGW YADSVTNRHE EKFSVDSYET DQMDTIYQCY NAVKMTKDGL TRVYVDRDGV NITVNLKPTG GLANGVRRYA SQTELYDAPG WLIWTYRTRT TVNCLITDMM AKSNSPFDFF VTTTGQTVEM SPFYDGKNKE TFHERADSFH VRTNYKIV | |
Greater than 90% as determined by SDS-PAGE. |
3783680 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |