missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr cytomegalovirus (HCMV) Glycoprotein B / gB Protein

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) cytomegalovirus (HCMV) Glycoprotein B / gB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7275-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077950

  • £715.00 / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

YP_401713.1
cytomegalovirus (HCMV) Glycoprotein B / gB Protein
Research Use Only
gB
Recombinant Protein
E. coli
QTPEQPAPPA TTVQPTATRQ QTSFPFRVCE LSSHGDLFRF SSDIQCPSFG TRENHTEGLL MVFKDNIIPY SFKVRSYTKI VTNILIYNGW YADSVTNRHE EKFSVDSYET DQMDTIYQCY NAVKMTKDGL TRVYVDRDGV NITVNLKPTG GLANGVRRYA SQTELYDAPG WLIWTYRTRT TVNCLITDMM AKSNSPFDFF VTTTGQTVEM SPFYDGKNKE TFHERADSFH VRTNYKIV
Greater than 90% as determined by SDS-PAGE.
3783680
100 μg
Not determined.
Partial (see sequence information for more details).
Epstein-Barr
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers