missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr DUTPase Protein

A DNA sequence encoding the Epstein-Barr virus (strain GD1) (HHV-4) (Human herpesvirus 4) DUTPase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7034-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017130

  • £1715.00 / 500µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

DUTPase Protein
Research Use Only
BLLF3
Recombinant Protein
E. coli
MEACPHIRYA FQNDKLLLQQ ASVGRLTLVN KTTILLRPMK TTTVDLGLYA RPPEGHGLML WGSTSRPVTS HVGIIDPGYT GELRLILQNQ RRYNSTLRPS ELKIHLAAFR YATPQMEEDK GPINHPQYPG DVGLDVSLPK DLALFPHQTV SVTLTVPPPS IPHHRPTIFG RSGLAMQGIL VKPCRWRRGG VDVSLTNFSD QTVFLNKYRR FCQLVYLHKH HLTSFYSPHS DAGVLGPRSL FRWASCAFEE VPSLAMGDSG LSEALEGRQG RGFGSSGQ
Greater than 90% as determined by SDS-PAGE.
500 μg
Not determined.
Full Length
Epstein-Barr
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers