missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr Latent membrane protein 1

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Latent membrane protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7054-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077230

  • £1930.00 / 500µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

YP_401631.1
Latent membrane protein 1
Research Use Only
LMP2
Recombinant Protein
Yeast
MGSLEMVPMG AGPPSPGGDP DGYDGGNNSQ YPSASGSSGN TPTPPNDEER ESNEEPPPPY EDPYWGNGDR HSDYQPLGTQ DQSLYLGLQH DGNDGLPPPP YSPRDDSSQH IYEEAGRGSM NPVCLPVIVA PYLFWLAAIA ASCFTAS
Greater than 80% as determined by SDS-PAGE.
17494212
500 μg
Not determined.
Partial (see sequence information for more details).
Epstein-Barr
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers