missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Equine IL-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1809-EL-020
282.50 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Equine IL-4 Protein is derived from E. coli. The Recombinant Equine IL-4 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
IL-4 | |
E. coli-derived equine IL-4 protein Lys26-Cys137 | |
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Recombinant |
12.6 kDa | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |