missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Feline Flt-3 Ligand/FLT3L Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 2467-FL-025
This item is not returnable.
View return policy
Description
Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse Flt-3. The ED50 for this effect is 0.100-1.00 ng/mL.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
Observed M.W.: 25-33 kDa, under reducing conditions. | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
493796 | |
Flt-3 Ligand | |
Mouse myeloma cell line,NS0-derived feline Flt-3 Ligand/FLT3L protein Ser27-Pro185,with a C-terminal 6-His tag | |
FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine | |
Recombinant |