missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 Integrase p31 Protein

A cDNA sequence encoding the HIV-1 Integrase p31 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12253-1mg

 View more versions of this product

Product Code. 15959838

  • £1320.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

HIV-1 Integrase p31 Protein
Research Use Only
Recombinant Protein
E. coli
1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
His
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers