missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 p66 pol Protein
A cDNA sequence encoding the HIV-1 p66 pol was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12259-10ug
This item is not returnable.
View return policy
Specifications
HIV-1 p66 pol Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The protein solution contains 30mM Tris pH 7, 0.15M NaCl and 0.2mM EDTA. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 90.0% as determined by HPLC analysis & SDS-PAGE. |