missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 TAT Clade-B Protein

A cDNA sequence encoding the HIV-1 TAT Clade-B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12263-10ug

 View more versions of this product

Product Code. 15959868

  • £155.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

HIV-1 TAT Clade-B Protein
Research Use Only
Recombinant Protein
E. coli
MEPVDPRLEP WKHPGSQPKT ACTNCYCKKC CFHCQVCFIT KALGISYGRK KRRQRRRPPQ GSQTHQVSLS KQPTSQSRGD PTGPKE
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
Lyophilized with 0.1% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers