missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV HTLV-1 Envelope Protein
A cDNA sequence encoding the HTLV-1 Envelope was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12353-500ug
This item is not returnable.
View return policy
Specifications
HIV HTLV-1 Envelope Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM Na-PO4 pH 6.0, 0.1% SDS and 1mM DTT. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
HTLV-1 Envelope protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |