missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HCV Fusion Protein
A cDNA sequence encoding the HCV Fusion was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12158-500ug
This item is not returnable.
View return policy
Specifications
HSV HCV Fusion Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile filtered solution containing 1x PBS and 50mM K2CO3. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |