missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human 14-3-3 zeta, HRP-conjugated Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ HRP2669

324.83 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18147760

  • £413.00 / 250µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Source: E. coli-derived, aa 1-245
Formulation: No Formulation Info in Bulk Inventory
Entrez Gene ID: 7534
Regulatory Status: RUO

Specifications

Specifications

Supplied as a 0.2μM filtered solution in phosphate-buffered saline (PBS) with stabilizers and preservatives.
250 μL
14-3-3 zeta
Human
7534
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP (Horseradish Peroxidase)
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers