missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 39S ribosomal protein L1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L1, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8132-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_064621.3 | |
39S ribosomal protein L1, mitochondrial Protein | |
Research Use Only | |
MRPL1 | |
Recombinant Protein | |
E. coli | |
KKTKKGAKEK TPDEKKDEIE KIKAYPYMEG EPEDDVYLKR LYPRQIYEVE KAVHLLKKFQ ILDFTSPKQS VYLDLTLDMA LGKKKNVEPF TSVLSLPYPF ASEINKVAVF TENASEVKIA EENGAAFAGG TSLIQKIWDD EIVADFYVAV PEIMPELNRL RKKLNKKYPK LSRNSIGRDI PKMLELFKNG HEIKVDEERE NFLQTKIATL DMSSDQIAAN LQAVINEVCR HRPLNLGPFV VRAFLRSSTS EGLLLKIDPL LPKEVKNEES EKED | |
Greater than 90% as determined by SDS-PAGE. |
65008 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |