missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 40S ribosomal protein S14 Protein

A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8471-ec-500ug

 View more versions of this product

Product Code. 16004801

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001020241.1
40S ribosomal protein S14 Protein
Research Use Only
RPS14
Recombinant Protein
E. coli
APRKGKEKKE EQVISLGPQV AEGENVFGVC HIFASFNDTF VHVTDLSGKE TICRVTGGMK VKADRDESSP YAAMLAAQDV AQRCKELGIT ALHIKLRATG GNRTKTPGPG AQSALRALAR SGMKIGRIED VTPIPSDSTR RKGGR
Greater than 90% as determined by SDS-PAGE.
6208
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers