missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 40S ribosomal protein S14 Protein
A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8471-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001020241.1 | |
40S ribosomal protein S14 Protein | |
Research Use Only | |
RPS14 | |
Recombinant Protein | |
E. coli | |
APRKGKEKKE EQVISLGPQV AEGENVFGVC HIFASFNDTF VHVTDLSGKE TICRVTGGMK VKADRDESSP YAAMLAAQDV AQRCKELGIT ALHIKLRATG GNRTKTPGPG AQSALRALAR SGMKIGRIED VTPIPSDSTR RKGGR | |
Greater than 90% as determined by SDS-PAGE. |
6208 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |