missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L11 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8307-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_000966.2 | |
60S ribosomal protein L11 Protein | |
Research Use Only | |
RPL11 | |
Recombinant Protein | |
E. coli | |
QDQGEKENPM RELRIRKLCL NICVGESGDR LTRAAKVLEQ LTGQTPVFSK ARYTVRSFGI RRNEKIAVHC TVRGAKAEEI LEKGLKVREY ELRKNNFSDT GNFGFGIQEH IDLGIKYDPS IGIYGLDFYV VLGRPGFSIA DKKRRTGCIG AKHRISKEEA MRWFQQKYDG IILPGK | |
Greater than 90% as determined by SDS-PAGE. |
6135 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |