missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACAA1 / PTHIO Protein
A DNA sequence encoding the Homo sapiens (Human) ACAA1 / PTHIO, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001123882.1 |
---|---|
Gene ID (Entrez) | 30 |
Name | ACAA1 / PTHIO Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966359
|
enQuireBio™
QP5602-ec-10ug |
10 μg | |||||||
15906369
|
enQuireBio™
QP5602-ec-50ug |
50 μg | |||||||
15956359
|
enQuireBio™
QP5602-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15986359
|
enQuireBio™
QP5602-ec-200ug |
200 μg | |||||||
15996359
|
enQuireBio™
QP5602-ec-500ug |
500 μg | |||||||
15976359
|
enQuireBio™
QP5602-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001123882.1 | |
ACAA1 / PTHIO Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
30 | |
Research Use Only | |
ACAA1 | |
Recombinant Protein | |
E. coli | |
LSGAPQASAA DVVVVHGRRT AICRAGRGGF KDTTPDELLS AVMTAVLKDV NLRPEQLGDI CVGNVLQPGA GAIMARIAQF LSDIPETVPL STVNRQCSSG LMS-QAVASI AGGIRNGSYD IGMACGITSE NVAERFGISR EKQDTFALAS QQKAARAQSK GCFQAEIVPV TTTVHDDKGT KRSITVTQDE GIRPSTTMEG LAKLKPAFKK DGSTTAGLTV SDVDIFEINE AFASQAAYCV EKLRLPPEKV NPLGGAVALG HPLGCTGARQ VITLLNELKR RGKRAYGVVS MCIGTGMGAA AVFEYPGN | |
Greater than 90% as determined by SDS-PAGE. |