missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Activated Coagulation Factor Xa, ACFP
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ ACFP1063
This item is not returnable.
View return policy
Description
The Recombinant Human Activated Coagulation Factor Xa, ACFP is derived from Sf 9 (baculovirus). The Recombinant Human Activated Coagulation Factor Xa, ACFP has been validated for the following applications: Enzyme Activity.Specifications
P00742 | |
2159 | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after reconstitution. | |
<1.0 EU per 1 μg of the protein by the LAL method. | |
Coagulation Factor X | |
Human | |
Reconstitute at 100 μg/mL in sterile 25 mM MES, 150 mM NaCl and 5 mM CaCl2, pH 6.0 | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Enzyme Activity | |
16.6 kDa & 30 kDa | |
Spodoptera frugiperda,Sf 9 (baculovirus)-derived human Coagulation Factor X protein Met1-Lys488,with a C-terminal 10-His tag Proform Factor X was expressed,purified,activated and further purified to yield Factor Xa. Produced in an animal component free process (ACFP). | |
RUO | |
F10 | |
Unconjugated | |
Recombinant | |
Lyophilized from a 0.2 μm filtered solution in MES, NaCl and CaCl2. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human Activated Coagulation Factor Xa, ACFP > Quantity: 25 μg
Spot an opportunity for improvement?Share a Content Correction