missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human ADAMTS4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 4307-AD-020
314.46 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human ADAMTS4 Protein is derived from CHO. The Recombinant Human ADAMTS4 Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2 μm filtered solution in Sodium Acetate, CaCl 2 and NaCl. | |
53 kDa | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
9507 | |
ADAMTS4 | |
Chinese Hamster Ovary cell line,CHO-derived human ADAMTS4 protein Phe213-Cys685,with a C-terminal 10-His tag | |
Recombinant Human ADAMTS4 is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag. | |
Recombinant |