missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Adenosine Kinase/ADK Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 8024-AK-025

273.86 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18177060

  • £348.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to phosphorylate Adenosine. The specific activity is >7 pmol/min/μg, as measured under the described conditions.
Specifications

Specifications

P55263
Supplied as a 0.2 μm filtered solution in Tris and NaCl.
M.W. (Observed): 43-45 kDa, reducing conditions; M.W. (theoretical): 41 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
ADK
Recombinant
ADK
Enzyme Activity
132
ADK
E. coli-derived human ADK protein Ala2-His362 with a C-terminal 6-His tag
<1.0 EU / 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers