missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AIP Protein
A cDNA sequence encoding the AIP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10988-1mg
This item is not returnable.
View return policy
Specifications
AIP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MADIIARLRE DGIQKRVIQE GRGELPDFQD GTKATFHYRT LHSDDEGTVL DDSRARGKPM ELIIGKKFKL PVWETIVCTM REGEIAQFLC DIKHVVLYPL VAKSLRNIAV GKDPLEGQRH CCGVAQMREH SSLGHADLDA LQQNPQPLIF HMEMLKVESP GTYQQDPWAM TDEEKAKAVP LIHQEGNRLY REGHVKEAAA KYYDAIACLK NLQMKEQPGS PEWIQLDKQI TPLLLNYCQC KLVVEEYYEV LDHCSSILNK YDDNVKAYFK RGKAHAAVWN AQEAQADFAK VLELDPALAP VVSRELRALE ARIRQKDEED KARFRGIFSH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
AIP protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |