missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AIP Protein
A cDNA sequence encoding the AIP was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | AIP Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975788
|
enQuireBio™
QP10988-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15965788
|
enQuireBio™
QP10988-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15955788
|
enQuireBio™
QP10988-1mg |
1 mg |
£2975.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
AIP Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
AIP protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MADIIARLRE DGIQKRVIQE GRGELPDFQD GTKATFHYRT LHSDDEGTVL DDSRARGKPM ELIIGKKFKL PVWETIVCTM REGEIAQFLC DIKHVVLYPL VAKSLRNIAV GKDPLEGQRH CCGVAQMREH SSLGHADLDA LQQNPQPLIF HMEMLKVESP GTYQQDPWAM TDEEKAKAVP LIHQEGNRLY REGHVKEAAA KYYDAIACLK NLQMKEQPGS PEWIQLDKQI TPLLLNYCQC KLVVEEYYEV LDHCSSILNK YDDNVKAYFK RGKAHAAVWN AQEAQADFAK VLELDPALAP VVSRELRALE ARIRQKDEED KARFRGIFSH | |
Greater than 90.0% as determined by SDS-PAGE. |