missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AKR1A1 Protein
A cDNA sequence encoding the AKR1A1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10994-20ug
This item is not returnable.
View return policy
Specifications
AKR1A1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAASCVLLHT GQKMPLIGLG TWKSEPGQVK AAVKYALSVG YRHIDCAAIY GNEPEIGEAL KEDVGPGKAV PREELFVTSK LWNTKHHPED VEPALRKTLA DLQLEYLDLY LMHWPYAFER GDNPFPKNAD GTICYDSTHY KETWKALEAL VAKGLVQALG LSNFNSRQID DILSVASVRP AVLQVECHPY LAQNELIAHC QARGLEVTAY SPLGSSDRAW RDPDEPVLLE PVVLALAEKY GRSPAQILL RWQVQRKVIC IPKSITPSRI LQNIKVFDFT FSPEEMKQLN ALNKNWRYIV PMLTVDGKRV PRDAGHPLYP FNDPY | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
AKR1A1 solution containing 20mM Tris pH 8.0, 50mM NaCl and 10% glycerol. |