missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALDH1A3 / RALDH3 Protein
A DNA sequence encoding the Homo sapiens (Human) ALDH1A3 / RALDH3, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_000684.2 |
---|---|
Gene ID (Entrez) | 220 |
Name | ALDH1A3 / RALDH3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15956669
|
enQuireBio™
QP5644-ec-10ug |
10 μg | N/A | ||||||
15996669
|
enQuireBio™
QP5644-ec-50ug |
50 μg | N/A | ||||||
15946669
|
enQuireBio™
QP5644-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15976669
|
enQuireBio™
QP5644-ec-200ug |
200 μg | N/A | ||||||
15986669
|
enQuireBio™
QP5644-ec-500ug |
500 μg | N/A | ||||||
15966669
|
enQuireBio™
QP5644-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000684.2 | |
ALDH1A3 / RALDH3 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
220 | |
Research Use Only | |
ALDH1A3 | |
Recombinant Protein | |
E. coli | |
ATANGAVENG QPDRKPPALP RPIRNLEVKF TKIFINNEWH ESKSGKKFAT CNPSTREQIC EVEEGDKPDV DKAVEAAQVA FQRGSPWRRL DALSRGRLLH QLADLVERDR ATLAALETMD TGKPFLHAFF IDLEGCIRTL RYFAGWADKI QGKTIPTDDN VVCFTRHEPI GVCGAITPWN FPLLMLVWKL APALCCGNTM VLKPAEQTPL TALYLGSLIK EAGFPPGVVN IVPGFGPTVG AAISSHPQIN KIAFTGSTEV GKLVKEAASR SNLKRVTLEL GGKNPCIVCA DADLDLAVEC AHQGVFFNQG QCCTAASRVF VEEQVYSEFV RRSVEYAKKR PVGDPFDVKT EQGPQIDQKQ FDKILELIES GKKEGAKLEC GGSAMEDKGL FIKPTVFSEV TDNMRIAKEE IFGPVQPILK FKSIEEVIKR ANSTDYGLTA AVFTKNLDKA LKLASALESG TVWINCYNAL YAQAPFGGFK MSGNGRELGE YALAEYTEVK TVTIKLGDKN P | |
Greater than 90% as determined by SDS-PAGE. |