missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANXA5 / Annexin 5 / Annexin A5 Protein
A DNA sequence encoding the Homo sapiens (Human) ANXA5 / Annexin 5 / Annexin A5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8982-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001145.1 | |
ANXA5 / Annexin 5 / Annexin A5 Protein | |
Research Use Only | |
ANXA5 | |
Recombinant Protein | |
Yeast | |
AQVLRGTVTD FPGFDERADA ETLRKAMKGL GTDEESILTL LTSRSNAQRQ EISAAFKTLF GRDLLDDLKS ELTGKFEKLI VALMKPSRLY DAYELKHALK GAGTNEKVLT EIIASRTPEE LRAIKQVYEE EYGSSLEDDV VGDTSGYYQR MLVVLLQANR DPDAGIDEAQ VEQDAQALFQ AGELKWGTDE EKFITIFGTR SVSHLRKVFD KYMTISGFQI EETIDRETSG NLEQLLLAVV KSIRSIPAYL AETLYYAMKG AGTDDHTLIR VMVSRSEIDL FNIRKEFRKN FATSLYSMIK GDTSGDYKKA LLLLCGEDD | |
Greater than 89.50% as determined by SDS-PAGE. |
308 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |