missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ASCC3 Protein

A DNA sequence encoding the Homo sapiens (Human) ASCC3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5683-ec-100ug

 View more versions of this product

Product Code. 15966979

  • £476.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001271200.1
ASCC3 Protein
Research Use Only
ASCC3
Recombinant Protein
E. coli
MALPRLTGAL RSFSNVTKQD NYNEEVADLK IKRSKLHEQV LDLGLTWKKI IKFLNEKLEK SKMQSINEDL KDILHAAKQI EVNCPFQKRR LDGKEEDEKM SRASDRFRGL R
Greater than 80% as determined by SDS-PAGE.
10973
100 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers