missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASCC3 Protein
A DNA sequence encoding the Homo sapiens (Human) ASCC3, was expressed in the hosts and tags indicated.
£183.90 - £2065.00
Specifications
Accession Number | NP_001271200.1 |
---|---|
Gene ID (Entrez) | 10973 |
Name | ASCC3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976979
|
enQuireBio™
QP5683-ec-10ug |
10 μg | |||||||
15916989
|
enQuireBio™
QP5683-ec-50ug |
50 μg | |||||||
15966979
|
enQuireBio™
QP5683-ec-100ug |
100 μg |
£476.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15996979
|
enQuireBio™
QP5683-ec-200ug |
200 μg | |||||||
15906989
|
enQuireBio™
QP5683-ec-500ug |
500 μg | |||||||
15986979
|
enQuireBio™
QP5683-ec-1mg |
1 mg |
£2065.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001271200.1 | |
ASCC3 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10973 | |
Research Use Only | |
ASCC3 | |
Recombinant Protein | |
E. coli | |
MALPRLTGAL RSFSNVTKQD NYNEEVADLK IKRSKLHEQV LDLGLTWKKI IKFLNEKLEK SKMQSINEDL KDILHAAKQI EVNCPFQKRR LDGKEEDEKM SRASDRFRGL R | |
Greater than 80% as determined by SDS-PAGE. |