missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP synthase subunit O, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ATP synthase subunit O, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8437-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_001688.1 | |
ATP synthase subunit O, mitochondrial Protein | |
Research Use Only | |
ATP5O | |
Recombinant Protein | |
E. coli | |
FAKLVRPPVQ VYGIEGRYAT ALYSAASKQN KLEQVEKELL RVAQILKEPK VAASVLNPYV KRSIKVKSLN DITAKERFSP LTTNLINLLA ENGRLSNTQG VVSAFSTMMS VHRGEVPCTV TSASPLEEAT LSELKTVLKS FLSQGQVLKL EAKTDPSILG GMIVRIGEKY VDMSVKTKIQ KLGRAMREIV | |
Greater than 90% as determined by SDS-PAGE. |
539 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |