missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP5D Protein
A DNA sequence encoding the Homo sapiens (Human) ATP5D, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5695-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001001975.1 | |
ATP5D Protein | |
Research Use Only | |
ATP5D | |
Recombinant Protein | |
E. coli | |
AEAAAAPAAA SGPNQMSFTF ASPTQVFFNG ANVRQVDVPT LTGAFGILAA HVPTLQVLRP GLVVVHAEDG TTSKYFVSSG SIAVNADSSV QLLAEEAVTL DMLDLGAAKA NLEKAQAELV GTADEATRAE IQIRIEANEA LVKALE | |
Greater than 90% as determined by SDS-PAGE. |
513 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |