missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human B3GAT3 Protein
A cDNA sequence encoding the B3GAT3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11118-1mg
This item is not returnable.
View return policy
Specifications
B3GAT3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQPCDCLP PLRAAAEQLR QKDLRISQLQ AELRRPPPAP AQPPEPEALP TIYVVTPTYA RLVQKAELVR LSQTLSLVPR LHWLLVEDAE GPTPLVSGLL AASGLLFTHL VVLTPKAQRL REGEPGWVHP RGVEQRNKAL DWLRGRGGAV GGEKDPPPPG TQGVVYFADD DNTYSRELFE EMRWTRGVSV WPVGLVGGLR FEGPQVQDGR VVGFHTAWEP SRPFPVDMAG FAVALPLLLD KPNAQFDSTA PRGHLESSLL SHLVDPKDLE PRAANCTRVL VWHTRTEKPK MKQEEQLQRQ GRGSDPAIEV | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
B3GAT3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |