missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human BAFF/BLyS/TNFSF13B Protein

Product Code. p-4719113
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18398387

Brand: R&D Systems™ 2149BF010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human BAFF/BLyS/TNFSF13B Protein is derived from NS0. The Recombinant Human BAFF/BLyS/TNFSF13B Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
Gene ID (Entrez) 10673
Molecular Weight (g/mol) 19.6 kDa
Name BAFF/BLyS/TNFSF13B
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived human BAFF/BLyS/TNFSF13B protein, (N-terminus) MHHHHHHHHHH GGGSGGGSGGGS IEGR Human BAFF (Ala134-Leu285) Accession &Num; Q9Y275 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.