missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCL7C Protein
A cDNA sequence encoding the BCL7C was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11147-2ug
This item is not returnable.
View return policy
Specifications
BCL7C Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGRTVR AETRSRAKDD IKKVMATIEK VRRWEKRWVT VGDTSLRIFK WVPVVDPQEE ERRRAGGGAE RSRGRERRGR GASPRGGGPL ILLDLNDENS NQSFHSEGSL QKGTEPSPGG TPQPSRPVSP AGPPEGVPEE AQPPRLGQER DPGGITAGST DEPPMLTKEE PVPELLEAEA PEAYPVFEPV PPVPEAAQGD TEDSEGAPPL KRICPNAPDP | |
Greater than 85% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BCL7C protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |