missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BCOADC-E2 Protein

A cDNA sequence encoding the BCOADC-E2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11148-10ug

 View more versions of this product

Product Code. 15956298

  • £155.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

BCOADC-E2 Protein
Research Use Only
Recombinant Protein
Baculovirus
DLST (0.91 mg/ml) is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl and 20% glycerol.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Greater than 75% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers