missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BDH2 Protein
A cDNA sequence encoding the BDH2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11150-1mg
This item is not returnable.
View return policy
Specifications
56898 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGRLDGKVII LTAAAQGIGQ AAALAFAREG AKVIATDINE SKLQELEKYP GIQTRVLDVT KKKQIDQFAN EVERLDVLFN VAGFVHHGTV LDCEEKDWDF SMNLNVRSMY LMIKAFLPKM LAQKSGNIIN MSSVASSVKG VVNRCVYSTT KAAVIGLTKS VAADFIQQGI RCNCVCPGTV DTPSLQERIQ ARGNPEEARN DFLKRQKTGR FATAEEIAML CVYLASDESA YVTGNPVIID GGWSL | |
Greater than 95% as determined by SDS-PAGE. |
BDH2 Protein | |
Research Use Only | |
BDH2 | |
Human | |
His | |
The BDH2 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 0.1M NaCl and 10% glycerol. |